accessibility menu, dialog, popup

UserName

CAS RN 144432-80-4

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-{[1,1'-biphenyl]-4-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 280.17
Molecular Formula: C18H21BO2
InChi Key: REDKQKNJWVIPIO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results