accessibility menu, dialog, popup

UserName

CAS RN 14442-12-7

OOON

CAS RN 14442-12-7

IUPAC Name: 3-phenyl-1,2-oxazole-5-carboxylate
Synonyms: 3-phenyl-1,2-oxazole-5-carboxylate
Molecular Weight (g/mol): 188.16
Molecular Formula: C10H6NO3
InChi Key: YGTFDJRCCBKLDM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)