accessibility menu, dialog, popup

UserName

H3CONCH3CH3

CAS RN 14433-76-2

IUPAC Name: N,N-dimethyldecanamide
Synonyms: dimethyl capramide
Molecular Weight (g/mol): 199.34
Molecular Formula: C12H25NO
InChi Key: HNXNKTMIVROLTK-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)N(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results