accessibility menu, dialog, popup

UserName

CAS RN 14405-45-9

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHIn

CAS RN 14405-45-9

IUPAC Name: indium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: indium acetylacetonate
Molecular Weight (g/mol): 412.15
Molecular Formula: C15H21InO6
InChi Key: CMGVNFXMZKDHDG-UHFFFAOYSA-N
SMILES: [In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results