CAS RN 14402-67-6
CAS RN 14402-67-6
IUPAC Name:
titanium(4+) dipotassium dihydrate dioxalate oxidandiide
Synonyms:
titanium(4+) dipotassium dihydrate dioxalate oxidandiide
Molecular Weight (g/mol):
354.13
Molecular Formula:
C4H4K2O11Ti
InChi Key:
GNRMKQMKBAJVLN-UHFFFAOYSA-J
SMILES:
O.O.[O--].[K+].[K+].[Ti+4].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
1
–
2
of
2
results
1
Potassium bis(oxalato)oxotitanate(IV) dihydrate
CAS: 14402-67-6 Molecular Formula: C4K2O9Ti Molecular Weight (g/mol): 318.10 MDL Number: MFCD00150592 InChI Key: UHWHMHPXHWHWPX-UHFFFAOYSA-J Synonym: Titanium potassium oxalate IUPAC Name: dipotassium oxotitaniumbis(ylium) dioxalate SMILES: [K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
2
Potassium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.997% (metals basis)
CAS: 14402-67-6 Molecular Formula: C4K2O9Ti Molecular Weight (g/mol): 318.10 MDL Number: MFCD00150592 InChI Key: UHWHMHPXHWHWPX-UHFFFAOYSA-J Synonym: Potassium titanyloxolate IUPAC Name: dipotassium oxotitaniumbis(ylium) dioxalate SMILES: [K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O