accessibility menu, dialog, popup

UserName

NH2OI

CAS RN 144-48-9

IUPAC Name: 2-iodoacetamide
Synonyms: iodoacetamide
Molecular Weight (g/mol): 184.96
Molecular Formula: C2H4INO
InChi Key: PGLTVOMIXTUURA-UHFFFAOYSA-N
SMILES: NC(=O)CI
Molecular Weight (g/mol) 
  • (7)
  • (8)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (5)
  • (3)
  • (7)
  • (8)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)