accessibility menu, dialog, popup

UserName

CH3CH3H3COOHNH2COOHHN

CAS RN 143979-15-1

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}pent-4-enoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-[(tert-butoxycarbonyl)amino]pent-4-enoic acid; dicha
Molecular Weight (g/mol): 396.57
Molecular Formula: C22H40N2O4
InChi Key: VMCGMPITVQIMGK-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CC=C)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)