accessibility menu, dialog, popup

UserName

CH3NNH2NClH3C

CAS RN 14394-61-7

IUPAC Name: 4-chloro-5,6-dimethylpyrimidin-2-amine
Synonyms: 4-chloro-5,6-dimethylpyrimidin-2-amine
Molecular Weight (g/mol): 157.6
Molecular Formula: C6H8ClN3
InChi Key: RRMVDLLCVGZDMH-UHFFFAOYSA-N
SMILES: CC1=NC(N)=NC(Cl)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results