accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHO

CAS RN 143900-44-1

IUPAC Name: tert-butyl 3-hydroxypiperidine-1-carboxylate
Synonyms: tert-butyl 3-hydroxypiperidine-1-carboxylate
Molecular Weight (g/mol): 201.27
Molecular Formula: C10H19NO3
InChi Key: UIJXHKXIOCDSEB-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(O)C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)