CAS RN 14384-67-9
IUPAC Name:
2-(4-phenyl-1,3-thiazol-2-yl)pyridine
Synonyms:
2-(4-phenyl-1,3-thiazol-2-yl)pyridine
Molecular Weight (g/mol):
238.31
Molecular Formula:
C14H10N2S
InChi Key:
ZRIPTGIQAZVFDP-UHFFFAOYSA-N
SMILES:
S1C=C(N=C1C1=CC=CC=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Phenyl-2-(2-pyridyl)thiazole, 97%, Thermo Scientific™
CAS: 14384-67-9 Molecular Formula: C14H10N2S Molecular Weight (g/mol): 238.308 MDL Number: MFCD09058229 InChI Key: ZRIPTGIQAZVFDP-UHFFFAOYSA-N Synonym: 4-phenyl-2-2-pyridyl thiazole,2-4-phenyl-1,3-thiazol-2-yl pyridine PubChem CID: 16832293 IUPAC Name: 4-phenyl-2-pyridin-2-yl-1,3-thiazole SMILES: C1=CC=C(C=C1)C2=CSC(=N2)C3=CC=CC=N3