accessibility menu, dialog, popup

UserName

CAS RN 143557-91-9

H3CCH3H3COONOH

CAS RN 143557-91-9

IUPAC Name: tert-butyl 3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
Synonyms: tert-butyl 3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
Molecular Weight (g/mol): 227.3
Molecular Formula: C12H21NO3
InChi Key: SEGZJJSZYOEABC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C2CCC1CC(O)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results