accessibility menu, dialog, popup

UserName

CAS RN 14347-78-5

CH3H3COOHO

CAS RN 14347-78-5

IUPAC Name: (2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Synonyms: solketal isopropylideneglycerol
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: RNVYQYLELCKWAN-UHFFFAOYNA-N
SMILES: CC1(C)OCC(CO)O1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)