accessibility menu, dialog, popup

UserName

CAS RN 143140-08-3

H3CNOH3CSiCH3H3CCH3

CAS RN 143140-08-3

IUPAC Name: (methoxymethyl)(1-phenylethyl)[(trimethylsilyl)methyl]amine
Synonyms: (methoxymethyl)(1-phenylethyl)[(trimethylsilyl)methyl]amine
Molecular Weight (g/mol): 251.45
Molecular Formula: C14H25NOSi
InChi Key: FRQGYHCIEIDEAD-UHFFFAOYNA-N
SMILES: COCN(C[Si](C)(C)C)C(C)C1=CC=CC=C1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)