accessibility menu, dialog, popup

UserName

CAS RN 1431329-77-9

CH3OFFFOCH3

CAS RN 1431329-77-9

IUPAC Name: 4-methoxy-3-methyl-5-(trifluoromethyl)benzaldehyde
Synonyms: 4-methoxy-3-methyl-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: UJHKXHXOLJJFDF-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=O)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results