accessibility menu, dialog, popup

UserName

HO

CAS RN 14300-33-5

IUPAC Name: dicyclopropylmethanol
Synonyms: dicyclopropyl carbinol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: PIXLZMHERIHLJL-UHFFFAOYSA-N
SMILES: OC(C1CC1)C1CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results