accessibility menu, dialog, popup

UserName

CAS RN 143-15-7

IUPAC Name: 1-bromododecane
Synonyms: 1-bromo-N-dodecane
Molecular Weight (g/mol): 249.24
Molecular Formula: C12H25Br
InChi Key: PBLNBZIONSLZBU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)