accessibility menu, dialog, popup

UserName

CAS RN 14297-34-8

OBr

CAS RN 14297-34-8

IUPAC Name: 2-(4-bromophenyl)furan
Synonyms: 2-(4-bromophenyl)furan
Molecular Weight (g/mol): 223.07
Molecular Formula: C10H7BrO
InChi Key: FBIKSVALROQWIY-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results