CAS RN 1429192-00-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Retro-2 cycl, MedChemExpress
MedChemExpress Retro-2 cycl (RN 1-001) is a dihydroquinazolinone (DHQZ) inhibitor of retrograde trafficking. Retro-2 cycl (RN 1-001) inhibits JCPyV and HPV16 pseudovirus with IC50s of 54 μM and 160 μM, respectively. Antiviral agent.