accessibility menu, dialog, popup

UserName

CAS RN 14285-68-8

ReReCCCCCCCCCCOOOOOOOOOO

CAS RN 14285-68-8

IUPAC Name: decakis(methanidylidyneoxidanium) dirhenium
Synonyms: dirhenium decacarbonyl
Molecular Weight (g/mol): 652.51
Molecular Formula: C10O10Re2
InChi Key: ZIZHEHXAMPQGEK-UHFFFAOYSA-N
SMILES: [Re].[Re].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)