CAS RN 1424799-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-SJ733, MedChemExpress
MedChemExpress (+)-SJ733 is an anti-malaria agent which can also inhibit Na+-ATPase PfATP4.