CAS RN 14228-73-0
CAS RN 14228-73-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,4-Cyclohexanedimethanol Diglycidyl Ether, TRC
CAS: 14228-73-0 Molecular Formula: C14H24O4 Molecular Weight (g/mol): 256.34 Synonym: 2,2'-[1,4-Cyclohexanediylbis(methyleneoxymethylene)]bis[oxirane],1,4-Bis(glycidoxymethyl)cyclohexane,1,4-Bis(hydroxymethyl)cyclohexane diglycidyl Ether,1,4-Bis[(2,3-epoxypropoxy)methyl]cyclohexane,1,4-Bis[(glycidyloxy)methyl]cyclohexane,1,4-Cyclohexanedimethanol diglycidyl Ether IUPAC Name: 2-[[4-(oxiran-2-ylmethoxymethyl)cyclohexyl]methoxymethyl]oxirane SMILES: C(OCC1CO1)C2CCC(COCC3CO3)CC2