CAS RN 1421373-98-9
CAS RN 1421373-98-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AZ-5104, MedChemExpress
MedChemExpress AZ-5104 is an active, demethylated metabolite of AZD 9291. AZ-5104 is an EGFR inhibitor with IC50s of 1, 6, 1, 25 and 7 nM for EGFRL858R/T790M, EGFRL858R, EGFRL861Q, EGFR and ErbB4, respectively.