missing translation for 'onlineSavingsMsg'
Learn More

AZ-5104, MedChemExpress

Product Code. p-200098130
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498456

Brand: Medchem Express HYB0793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZ-5104 is an active, demethylated metabolite of AZD 9291. AZ-5104 is an EGFR inhibitor with IC50s of 1, 6, 1, 25 and 7 nM for EGFRL858R/T790M, EGFRL858R, EGFRL861Q, EGFR and ErbB4, respectively.

AZ-5104 is an active, demethylated metabolite of AZD 9291. AZ-5104 is an EGFR inhibitor with IC50s of 1, 6, 1, 25 and 7 nM for EGFRL858R/T790M, EGFRL858R, EGFRL861Q, EGFR and ErbB4, respectively.

Specifications

Chemical Name or Material AZ-5104
CAS 1421373-98-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H31N7O2
Quantity 100 mg
Solubility Information DMSO : ≥ 28 mg/mL (57.66 mM)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CNC4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C)=O
Molecular Weight (g/mol) 485.58
Formula Weight 485.58
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.