accessibility menu, dialog, popup

UserName

CAS RN 1421263-42-4

H3COSONHN

CAS RN 1421263-42-4

IUPAC Name: 5-(5-methylfuran-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 5-(5-methylfuran-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 232.26
Molecular Formula: C11H8N2O2S
InChi Key: PMVYUYYRYHAMLD-UHFFFAOYSA-N
SMILES: CC1=CC=C(O1)C1=CSC2=C1C(=O)NC=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results