accessibility menu, dialog, popup

UserName

CH3H3CNNNBrH3C(?)

CAS RN 1421261-26-8

IUPAC Name: 3-(4-bromophenyl)-4-(2,6-dimethylphenyl)-5-methyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-4-(2,6-dimethylphenyl)-5-methyl-1,2,4-triazole
Molecular Weight (g/mol): 342.24
Molecular Formula: C17H16BrN3
InChi Key: BRDRVRSFOWPFQM-UHFFFAOYSA-N
SMILES: CC1=NN=C(N1C1=C(C)C=CC=C1C)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results