accessibility menu, dialog, popup

UserName

CAS RN 142-29-0

CAS RN 142-29-0

IUPAC Name: cyclopentene
Synonyms: cyclopentene
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: LPIQUOYDBNQMRZ-UHFFFAOYSA-N
SMILES: C1CC=CC1
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)