accessibility menu, dialog, popup

UserName

CAS RN 142-03-0

H3COOAlOHOOCH3

CAS RN 142-03-0

IUPAC Name: bis(acetyloxy)alumanyl hydrate
Synonyms: bis(acetyloxy)alumanyl hydrate
Molecular Weight (g/mol): 163.09
Molecular Formula: C4H8AlO5
InChi Key: RHPIJWYTYJJCFU-UHFFFAOYSA-L
SMILES: O.CC(=O)O[Al]OC(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)