accessibility menu, dialog, popup

UserName

CH3NH3CH3COOCH3

CAS RN 14186-50-6

IUPAC Name: methyl 1,2,5-trimethyl-1H-pyrrole-3-carboxylate
Synonyms: methyl 1,2,5-trimethylpyrrole-3-carboxylate
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: VRUCCWQGKPXNMI-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)N(C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results