accessibility menu, dialog, popup

UserName

CAS RN 141478-68-4

FFFSOOOFFFSOOOFFFSOOOTm

CAS RN 141478-68-4

IUPAC Name: thulium(3+) tritrifluoromethanesulfonate
Synonyms: thulium(3+) tritriflate
Molecular Weight (g/mol): 616.12
Molecular Formula: C3F9O9S3Tm
InChi Key: PBASUZORNBYVFM-UHFFFAOYSA-K
SMILES: [Tm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results