accessibility menu, dialog, popup

UserName

ClClNN

CAS RN 141-30-0

IUPAC Name: 3,6-dichloropyridazine
Synonyms: pyridazine, 3,6-dichloro-
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: GUSWJGOYDXFJSI-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)N=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)