accessibility menu, dialog, popup

UserName

CAS RN 1408074-51-0

NHOOOOHHO

CAS RN 1408074-51-0

IUPAC Name: 1-oxa-6-azaspiro[3.4]octane; oxalic acid
Synonyms: 1-oxa-6-azaspiro[3.4]octane; oxalic acid
Molecular Weight (g/mol): 203.19
Molecular Formula: C8H13NO5
InChi Key: ZKTJFOHJBNDMDS-UHFFFAOYNA-N
SMILES: OC(=O)C(O)=O.C1CC2(CCNC2)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results