accessibility menu, dialog, popup

UserName

CAS RN 140712-79-4

N(R)NONHN(R)NHO(S)OOOCH3OH3C

CAS RN 140712-79-4

IUPAC Name: N-{9-[(2R,4S,5R)-4-[bis(4-methoxyphenyl)(phenyl)methoxy]-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
Synonyms: N-{9-[(2R,4S,5R)-4-[bis(4-methoxyphenyl)(phenyl)methoxy]-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl}benzamide
Molecular Weight (g/mol): 657.73
Molecular Formula: C38H35N5O6
InChi Key: LFXBQKFIXWICJR-WIHCDAFUSA-N
SMILES: COC1=CC=C(C=C1)C(O[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results