accessibility menu, dialog, popup

UserName

NNFClBFFFFBFFFF

CAS RN 140681-55-6

IUPAC Name: 1-(chloromethyl)-4-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium; bis(tetrafluoroboranuide)
Synonyms: selectfluor
Molecular Weight (g/mol): 354.26
Molecular Formula: C7H14B2ClF9N2
InChi Key: TXRPHPUGYLSHCX-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results