accessibility menu, dialog, popup

UserName

NH(S)HN(S)(S)NN(S)(R)HNNHONH2(S)HN(S)ONH2CH3H3COOH2NOO(R)HOCH3OHHN(S)HNH2NO(S)ONH(R)(S)CH3OHH2NONHOH3CCH3HHOHSOHOOOHSOHOO

CAS RN 1405-20-5

IUPAC Name: 4-amino-N-{1-[(3-amino-1-{[6,9,18-tris(2-aminoethyl)-15-benzyl-3-(2-hydroxyethyl)-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)carbamoyl]-3-hydroxypropyl}-2-formamidobutanamide; sulfuric acid
Synonyms: polymyxin B
Molecular Weight (g/mol): 1189.36
Molecular Formula: C48H84N16O17S
InChi Key: HNDFYNOVSOOGDU-UHFFFAOYNA-N
SMILES: OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (3)
Physical Form 
  • (9)
  • (1)
Color 
  • (8)
Type 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (9)
  • (1)
  • (8)
  • (7)
  • (1)
  • (3)
  • (1)
  • (1)