accessibility menu, dialog, popup

UserName

OClOOOOClOOOOClOOOTm

CAS RN 14014-10-9

IUPAC Name: thulium(3+) triperchlorate
Synonyms: thulium(3+) triperchlorate
Molecular Weight (g/mol): 467.27
Molecular Formula: Cl3O12Tm
InChi Key: JNNWMYZIHRZEKX-UHFFFAOYSA-K
SMILES: [Tm+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results