accessibility menu, dialog, popup

UserName

CH3NHNNOO

CAS RN 14003-66-8

IUPAC Name: 5-methyl-4-nitro-1H-imidazole
Synonyms: 4-methyl-5-nitro-3H-imidazole
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: WSYOWIMKNNMEMZ-UHFFFAOYSA-N
SMILES: CC1=C(N=CN1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)