accessibility menu, dialog, popup

UserName

ClO

CAS RN 14002-51-8

IUPAC Name: [1,1'-biphenyl]-4-carbonyl chloride
Synonyms: [1,1'-biphenyl]-4-carbonyl chloride
Molecular Weight (g/mol): 216.66
Molecular Formula: C13H9ClO
InChi Key: JPVUWCPKMYXOKW-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results