accessibility menu, dialog, popup

UserName

H3CCH3(R)PNH2

CAS RN 1400149-69-0

IUPAC Name: (2-amino-3-methylbutyl)diphenylphosphane
Synonyms: (2-amino-3-methylbutyl)diphenylphosphane
Molecular Weight (g/mol): 271.34
Molecular Formula: C17H22NP
InChi Key: ZZLCXURCZWQECA-UHFFFAOYNA-N
SMILES: CC(C)C(N)CP(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results