accessibility menu, dialog, popup

UserName

CAS RN 140-88-5

IUPAC Name: ethyl prop-2-enoate
Synonyms: ethyl acrylate polyethylacrylate
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: JIGUQPWFLRLWPJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C=C
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)