CAS RN 13967-90-3
IUPAC Name:
barium(2+) dibromate
Synonyms:
barium(2+) dibromate
Molecular Weight (g/mol):
393.13
Molecular Formula:
BaBr2O6
InChi Key:
VEASZGAADGZARC-UHFFFAOYSA-L
SMILES:
[Ba++].[O-][Br](=O)=O.[O-][Br](=O)=O
1
–
1
of
1
results
1
Barium bromate, 97%
CAS: 13967-90-3 Molecular Formula: BaBr2O6 Molecular Weight (g/mol): 393.129 MDL Number: MFCD00014177 InChI Key: VEASZGAADGZARC-UHFFFAOYSA-L Synonym: barium bromate,bromic acid, barium salt,unii-5qk4064y0r,barium 2+ dibromate,acmc-1bumj,ba.2bro3,bromic acid, barium salt 2:1,barium bromate un2719 oxidizer PubChem CID: 61706 IUPAC Name: barium(2+);dibromate SMILES: [O-]Br(=O)=O.[O-]Br(=O)=O.[Ba+2]