accessibility menu, dialog, popup

UserName

CAS RN 13958-93-5

NClOOHCl

CAS RN 13958-93-5

IUPAC Name: 3,5-dichloropyridine-4-carboxylic acid
Synonyms: 3,5-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: BUQPTOSHKHYHHB-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=NC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results