CAS RN 13943-58-3
IUPAC Name:
tetrapotassium ironhexacarbonitrile
Synonyms:
potassium ferrocyanide
Molecular Weight (g/mol):
374.39
Molecular Formula:
C6H6FeK4N6
InChi Key:
FIBBHPJORRWWPX-UHFFFAOYSA-N
SMILES:
[K+].[K+].[K+].[K+].[Fe].C#N.C#N.C#N.C#N.C#N.C#N
1
–
2
of
2
results
2
Redox Buffer, 220mV, Reagecon™
Used for testing, calibration and verification of redox/ORP sensors. Redox Buffer, 220mV, Reagecon™ is a ready to use solution of iron salts, including potassium ferrocyanide and tripotassium hexacyanoferrate, for use with reactions with a redox potential of 220mV.