CAS RN 1393813-41-6
CAS RN 1393813-41-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Propylsulfamide Potassium Salt, TRC
CAS: 1393813-41-6 Molecular Formula: C3H10KN2O2S Molecular Weight (g/mol): 177.29 Synonym: Propylsulfamide potassium IUPAC Name: potassium propyl(sulfamoyl)amide SMILES: NS([N-]CCC)(=O)=O.[K+]