CAS RN 1391053-88-5
CAS RN 1391053-88-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Tyrosol 3-Sulfate Sodium Salt, TRC
CAS: 1391053-88-5 Molecular Formula: C8H9NaO6S Molecular Weight (g/mol): 256.21 Synonym: 4-(2-Hydroxyethyl)-1,2-benzenediol 2-(Hydrogen Sulfate) Sodium; IUPAC Name: sodium 2-hydroxy-5-(2-hydroxyethyl)phenyl sulfate SMILES: OC1=CC=C(CCO)C=C1OS(=O)(O[Na])=O