accessibility menu, dialog, popup

UserName

CAS RN 139062-96-7

H3CONHNCH3

CAS RN 139062-96-7

IUPAC Name: 1-[4-(methylamino)piperidin-1-yl]ethan-1-one
Synonyms: 1-[4-(methylamino)piperidin-1-yl]ethanone
Molecular Weight (g/mol): 156.23
Molecular Formula: C8H16N2O
InChi Key: RSEPODZAQBVPOS-UHFFFAOYSA-N
SMILES: CNC1CCN(CC1)C(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results