CAS RN 138872-73-8
CAS RN 138872-73-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deschloro Florfenicol, TRC
CAS: 138872-73-8 Molecular Formula: C12 H15 Cl F N O4 S Molecular Weight (g/mol): 323.77 Synonym: Acetamide, 2-chloro-N-[(1S,2R)-1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]-,Acetamide, 2-chloro-N-[1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]-, [R-(R*,S*)]-,2-Chloro-N-[(1S,2R)-1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]acetamide,Florfenicol-deschloro IUPAC Name: 2-chloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide SMILES: CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@@H](CF)NC(=O)CCl