accessibility menu, dialog, popup

UserName

OOHNHOO

CAS RN 138774-93-3

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-1-yl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(naphthalen-1-yl)propanoic acid
Molecular Weight (g/mol): 437.5
Molecular Formula: C28H23NO4
InChi Key: ORWNVJDLEMVDLV-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)