accessibility menu, dialog, popup

UserName

CAS RN 138650-24-5

OOOHNH2

CAS RN 138650-24-5

IUPAC Name: 3-aminooxetane-3-carboxylic acid
Synonyms: 3-aminooxetane-3-carboxylic acid
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: LXRNYRAHXDUNKR-UHFFFAOYSA-N
SMILES: NC1(COC1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results