accessibility menu, dialog, popup

UserName

CAS RN 138647-49-1

CH3H3CCH3OONOSOOFFF

CAS RN 138647-49-1

IUPAC Name: tert-butyl 4-(trifluoromethanesulfonyloxy)-1,2,3,6-tetrahydropyridine-1-carboxylate
Synonyms: tert-butyl 4-(trifluoromethanesulfonyloxy)-3,6-dihydro-2H-pyridine-1-carboxylate
Molecular Weight (g/mol): 331.31
Molecular Formula: C11H16F3NO5S
InChi Key: WUBVEMGCQRSBBT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(OS(=O)(=O)C(F)(F)F)=CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results