CAS RN 1384516-10-2
CAS RN 1384516-10-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(4-Chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine, TRC
CAS: 1384516-10-2 Molecular Formula: C12H11ClN2O2S Molecular Weight (g/mol): 282.75 Synonym: N-[(4-Chlorophenyl)methyl]-5-nitro-2-thiophenemethanamine (ACI), N-(4-Chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine IUPAC Name: N-(4-chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine SMILES: O=[N+](C1=CC=C(CNCC2=CC=C(Cl)C=C2)S1)[O-]